1-(2,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
Dodavatel: Apollo Scientific
OR32028-1G
OR32028-5G
APOSOR32028-1GEA
4630
CZK
APOSOR32028-1G
APOSOR32028-5G
1-(2,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
1-(2,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Vzorec:
C₁₃H₉Cl₂NO₃ MW: 298,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00140355 CAS číslo: 242797-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...