2-(1H-1,2,4-Triazol-1-yl)acetonitrile 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(1,2,4-Triazol-1-yl)acetonitrile
OR32027-1G
OR32027-5G
APOSOR32027-1GEA
1820
CZK
APOSOR32027-1G
APOSOR32027-5G
2-(1H-1,2,4-Triazol-1-yl)acetonitrile 95%
2-(1H-1,2,4-Triazol-1-yl)acetonitrile
Vzorec:
C₄H₄N₄ MW: 108,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792910 CAS číslo: 81606-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...