Tisk…

Ethyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate 95%

Dodavatel: Apollo Scientific

OR32026-1G OR32026-5G OR32026-25G
APOSOR32026-1GEA 3480 CZK
APOSOR32026-1G APOSOR32026-5G APOSOR32026-25G
Ethyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate 95%
Ethyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate
Vzorec: C₈H₁₃N₃O₂
MW: 183,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00202105
CAS číslo: 100159-13-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR