Ethyl-2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR32020-1G
OR32020-5G
OR32020-25G
APOSOR32020-1GEA
5130
CZK
APOSOR32020-1G
APOSOR32020-5G
APOSOR32020-25G
Ethyl-2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate 95%
Ethyl-2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Vzorec:
C₁₂H₁₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00139511 CAS číslo: 241146-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...