4-Methylbenzyl mercaptan 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
p-Xylene-α-thiol
OR31845-25ML
OR31845-5ML
APOSOR31845-25MLEA
13460
CZK
APOSOR31845-25ML
APOSOR31845-5ML
4-Methylbenzyl mercaptan 97%
4-Methylbenzyl mercaptan
Vzorec:
C₈H₁₀S MW: 138,23 g/mol Bod tání: 116…118/35mm °C Hustota: 1,023 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039652 CAS číslo: 4498-99-1 EINECS: 224-796-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...