Tisk…

(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(dicyclohexylphosphino)ferrocene, [(R)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyl]di-tert-butylphosphine

OR31838-1G OR31838-250MG OR31838-5G
APOSOR31838-1GEA 1080 CZK
APOSOR31838-1G APOSOR31838-250MG APOSOR31838-5G
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine 95%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine
Vzorec: C₃₂H₅₂FeP₂
MW: 554,5500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08543435
CAS číslo: 158923-11-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR