Tisk…

Ethyl-5-amino-1-benzyl-1H-imidazole-4-carboxylate 95+%

Dodavatel: Apollo Scientific

OR318101-1G OR318101-10G OR318101-5G
APOSOR318101-1GEA 7340 CZK
APOSOR318101-1G APOSOR318101-10G APOSOR318101-5G
Ethyl-5-amino-1-benzyl-1H-imidazole-4-carboxylate 95+%
Ethyl-5-amino-1-benzyl-1H-imidazole-4-carboxylate
Vzorec: C₁₃H₁₅N₃O₂
MW: 245,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03406879
CAS číslo: 68462-61-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR