Tisk…

Methyl-3-amino-5-methylthiophene-2-carboxylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 3-amino-5-methylthiophene-2-carboxylate

OR318100-1G OR318100-5G OR318100-10G
APOSOR318100-1GEA 1910 CZK
APOSOR318100-1G APOSOR318100-5G APOSOR318100-10G
Methyl-3-amino-5-methylthiophene-2-carboxylate 97%
Methyl-3-amino-5-methylthiophene-2-carboxylate
Vzorec: C₇H₉NO₂S
MW: 171,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130095
CAS číslo: 76575-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR