(1-(Pyridin-2-yl)cyclohexyl)methanamine tosylate salt 95+%
Dodavatel: Apollo Scientific
OR318090-1G
OR318090-10G
OR318090-5G
APOSOR318090-1GEA
13860
CZK
APOSOR318090-1G
APOSOR318090-10G
APOSOR318090-5G
(1-(Pyridin-2-yl)cyclohexyl)methanamine tosylate salt 95+%
(1-(Pyridin-2-yl)cyclohexyl)methanamine tosylate salt
Vzorec:
C₁₉H₂₆N₂O₃S MW: 362,49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226765 CAS číslo: 1170797-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...