2-Cyclopropylbenzoic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR318088-1G
OR318088-10G
OR318088-5G
APOSOR318088-1GEA
3480
CZK
APOSOR318088-1G
APOSOR318088-10G
APOSOR318088-5G
2-Cyclopropylbenzoic acid 95+%
2-Cyclopropylbenzoic acid
Vzorec:
C₁₀H₁₀O₂ Bod tání: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD01550965 CAS číslo: 3158-74-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...