4-(2-Hydrazinylethyl)morpholine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR318074-1G
OR318074-250MG
APOSOR318074-1GEA
3710
CZK
APOSOR318074-1G
APOSOR318074-250MG
4-(2-Hydrazinylethyl)morpholine 95%
4-(2-Hydrazinylethyl)morpholine
Vzorec:
C₆H₁₅N₃O Bod varu: 80 °C/0,075mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD07786279 CAS číslo: 2154-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...