Tisk…

tert-Butyl piperidine-3-carboxylate 95+%

Dodavatel: Apollo Scientific

OR318067-1G OR318067-5G
APOSOR318067-1GEA 12570 CZK
APOSOR318067-1G APOSOR318067-5G
tert-Butyl piperidine-3-carboxylate 95+%
tert-Butyl piperidine-3-carboxylate
Vzorec: C₁₀H₁₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD11041017
CAS číslo: 301180-05-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR