1-(Pyridin-2-yl)cyclohexanecarbonitrile 95+%
Dodavatel: Apollo Scientific
OR318060-1G
OR318060-5G
APOSOR318060-1GEA
9230
CZK
APOSOR318060-1G
APOSOR318060-5G
1-(Pyridin-2-yl)cyclohexanecarbonitrile 95+%
1-(Pyridin-2-yl)cyclohexanecarbonitrile
Vzorec:
C₁₂H₁₄N₂ Storage Temperature: Ambient |
MDL Number:
MFCD11052314 CAS číslo: 204067-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...