Tisk…

N-(Pyridin-3-ylmethyl)ethanamine 95+%

Dodavatel: Apollo Scientific

Synonyms: N-Ethyl-N-(3-pyridylmethyl)amine

OR318059-10G OR318059-5G
APOSOR318059-10GEA 2570 CZK
APOSOR318059-10G APOSOR318059-5G
N-(Pyridin-3-ylmethyl)ethanamine 95+%
N-(Pyridin-3-ylmethyl)ethanamine
Vzorec: C₈H₁₂N₂
MW: 136,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04557869
CAS číslo: 3000-75-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR