2-(2-Mercaptoethyl)isoindoline-1,3-dione 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(2-Sulphanylethyl)phthalimide
OR318057-1G
OR318057-10G
OR318057-5G
APOSOR318057-1GEA
14320
CZK
APOSOR318057-1G
APOSOR318057-10G
APOSOR318057-5G
2-(2-Mercaptoethyl)isoindoline-1,3-dione 95+%
2-(2-Mercaptoethyl)isoindoline-1,3-dione
Vzorec:
C₁₀H₉NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD00023091 CAS číslo: 4490-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...