Ethyl-2-(3-iodophenoxy)acetate 95+%
Dodavatel: Apollo Scientific
OR318049-1G
OR318049-10G
OR318049-5G
APOSOR318049-1GEA
7770
CZK
APOSOR318049-1G
APOSOR318049-10G
APOSOR318049-5G
Ethyl-2-(3-iodophenoxy)acetate 95+%
Ethyl-2-(3-iodophenoxy)acetate
Vzorec:
C₁₀H₁₁IO₃ MW: 306,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659701 CAS číslo: 90888-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...