Ethyl-2-m-tolylpyrimidine-5-carboxylate 95+%
Dodavatel: Apollo Scientific
OR318042-1G
OR318042-5G
APOSOR318042-1GEA
16870
CZK
APOSOR318042-1G
APOSOR318042-5G
Ethyl-2-m-tolylpyrimidine-5-carboxylate 95+%
Ethyl-2-m-tolylpyrimidine-5-carboxylate
Vzorec:
C₁₄H₁₄N₂O₂ MW: 242,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09863220 CAS číslo: 954227-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...