2,6-Di(pyridin-4-yl)pyrimidin-4-ol 95+%
Dodavatel: Apollo Scientific
OR318040-1G
OR318040-5G
OR318040-10G
APOSOR318040-1GEA
14120
CZK
APOSOR318040-1G
APOSOR318040-5G
APOSOR318040-10G
2,6-Di(pyridin-4-yl)pyrimidin-4-ol 95+%
2,6-Di(pyridin-4-yl)pyrimidin-4-ol
Vzorec:
C₁₄H₁₀N₄O Storage Temperature: Ambient |
MDL Number:
MFCD10688280 CAS číslo: 14757-05-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...