Imidazo[2,1-b]benzothiazole-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR318037-1G
OR318037-10G
OR318037-5G
APOSOR318037-1GEA
5560
CZK
APOSOR318037-1G
APOSOR318037-10G
APOSOR318037-5G
Imidazo[2,1-b]benzothiazole-2-carboxylic acid 95%
Imidazo[2,1-b]benzothiazole-2-carboxylic acid
Vzorec:
C₁₀H₆N₂O₂S MW: 218,24 g/mol Bod tání: 256…259 °C Storage Temperature: Ambient |
MDL Number:
MFCD00203011 CAS číslo: 64951-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...