Tisk…

5-Cyclopropyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one 95+%

Dodavatel: Apollo Scientific

OR318036-1G OR318036-5G OR318036-10G
APOSOR318036-1GEA 11680 CZK
APOSOR318036-1G APOSOR318036-5G APOSOR318036-10G
5-Cyclopropyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one 95+%
5-Cyclopropyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one
Vzorec: C₇H₈N₂OS
Storage Temperature: Ambient
MDL Number: MFCD10688279
CAS číslo: 59698-21-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR