Tisk…

3,5-Bis(methylsulphonyl)benzylamine hydrochloride 95+%

Dodavatel: Apollo Scientific

OR318026-1G OR318026-10G OR318026-5G
APOSOR318026-1GEA 9060 CZK
APOSOR318026-1G APOSOR318026-10G APOSOR318026-5G
3,5-Bis(methylsulphonyl)benzylamine hydrochloride 95+%
3,5-Bis(methylsulphonyl)benzylamine hydrochloride
Vzorec: C₉H₁₄ClNO₄S₂
Storage Temperature: Ambient
MDL Number: MFCD10688274
CAS číslo: 1171787-85-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR