Tisk…

3-Hydroxythiophene-2-carboxamide 95+%

Dodavatel: Apollo Scientific
Varování

OR318023-1G OR318023-10G OR318023-5G
APOSOR318023-1GEA 5560 CZK
APOSOR318023-1G APOSOR318023-10G APOSOR318023-5G
3-Hydroxythiophene-2-carboxamide 95+%
3-Hydroxythiophene-2-carboxamide
Vzorec: C₅H₅NO₂S
Storage Temperature: Ambient
MDL Number: MFCD09057756
CAS číslo: 57059-24-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR