Tisk…

tert-Butyl-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-2-carboxylate 95+%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 6-hydroxy-3,4-dihydroisoquinoline-2(1H)-carboxylate

OR318022-1G OR318022-10G OR318022-5G
APOSOR318022-1GEA 3150 CZK
APOSOR318022-1G APOSOR318022-10G APOSOR318022-5G
tert-Butyl-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-2-carboxylate 95+%
tert-Butyl-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
Vzorec: C₁₄H₁₉NO₃
MW: 249,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275034
CAS číslo: 158984-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR