Tisk…

6-Aminochromen-2-one 95+%

Dodavatel: Apollo Scientific

OR318015-1G OR318015-25G OR318015-5G
APOSOR318015-1GEA 2570 CZK
APOSOR318015-1G APOSOR318015-25G APOSOR318015-5G
6-Aminochromen-2-one 95+%
6-Aminochromen-2-one
Vzorec: C₉H₇NO₂
MW: 161.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115435
CAS číslo: 14415-44-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR