Tisk…

N-Benzoylethylenediamine hydrochloride 95+%

Dodavatel: Apollo Scientific

OR318004-1G OR318004-5G
APOSOR318004-1GEA 15150 CZK
APOSOR318004-1G APOSOR318004-5G
N-Benzoylethylenediamine hydrochloride 95+%
N-Benzoylethylenediamine hydrochloride
Vzorec: C₉H₁₃ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD10511533
CAS číslo: 1502-45-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR