Tisk…
3-(1,2,3,4-Tetrahydroisoquinoline-2-yl)propionic acid hydrochloride 95+%
Dodavatel: Apollo Scientific
OR318002-1G OR318002-5G
APOSOR318002-1GEA 13860 CZK
APOSOR318002-1G APOSOR318002-5G
3-(1,2,3,4-Tetrahydroisoquinoline-2-yl)propionic acid hydrochloride 95+%
3-(1,2,3,4-Tetrahydroisoquinoline-2-yl)propionic acid hydrochloride
Vzorec: C₁₂H₁₆ClNO₂
MW: 241.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10688268
CAS číslo: 846576-15-6