1-Bromo-5-nitronaphthalene 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR3179-100G
OR3179-25G
OR3179-5G
APOSOR3179-100GEA
5230
CZK
APOSOR3179-100G
APOSOR3179-25G
APOSOR3179-5G
1-Bromo-5-nitronaphthalene 96%
1-Bromo-5-nitronaphthalene
Vzorec:
C₁₀H₆BrNO₂ MW: 252.0641 g/mol Bod tání: 116…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156397 CAS číslo: 5328-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...