Tisk…

4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine, 4-Chloro-7-(p-toluenesulphonyl)-7H-pyrrolo[2,3-d]pyrimidine

OR317249-10G OR317249-1G OR317249-5G
APOSOR317249-10GEA 16340 CZK
APOSOR317249-10G APOSOR317249-1G APOSOR317249-5G
4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine
4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine
Vzorec: C₁₃H₁₀ClN₃O₂S
MW: 307,7600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907939
CAS číslo: 479633-63-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR