1-Methylcyclobutanecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR317127-1G
OR317127-5G
APOSOR317127-1GEA
1870
CZK
APOSOR317127-1G
APOSOR317127-5G
1-Methylcyclobutanecarboxylic acid
1-Methylcyclobutanecarboxylic acid
Vzorec:
C₆H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD15527494 CAS číslo: 32936-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...