3,3-Dimethylcyclobutanecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR317125-1G
OR317125-250MG
APOSOR317125-1GEA
4470
CZK
APOSOR317125-1G
APOSOR317125-250MG
3,3-Dimethylcyclobutanecarboxylic acid
3,3-Dimethylcyclobutanecarboxylic acid
Vzorec:
C₇H₁₂O₂ Bod varu: 105…106 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01734205 CAS číslo: 34970-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...