Tisk…

3-(Boc-amino)oxetane-3-methanol 97%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl (3-(hydroxymethyl)oxetan-3-yl)carbamate

OR317049-1G OR317049-500MG
APOSOR317049-1GEA 19840 CZK
APOSOR317049-1G APOSOR317049-500MG
3-(Boc-amino)oxetane-3-methanol 97%
3-(Boc-amino)oxetane-3-methanol
Vzorec: C₉H₁₇NO₄
MW: 203,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415250
CAS číslo: 1363382-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR