4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR317011-1G
OR317011-5G
APOSOR317011-1GEA
3110
CZK
APOSOR317011-1G
APOSOR317011-5G
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
Vzorec:
C₇H₃ClN₄ Storage Temperature: Ambient |
MDL Number:
MFCD12406095 CAS číslo: 24391-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...