4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR317010-1G
OR317010-5G
APOSOR317010-1GEA
2940
CZK
APOSOR317010-1G
APOSOR317010-5G
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde
Vzorec:
C₇H₄ClN₃O Storage Temperature: Ambient |
MDL Number:
MFCD12406099 CAS číslo: 908287-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...