5'-Bromo-2'-chloroacetophenone 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(5-Bromo-2-chlorophenyl)ethanone
OR3169-1G
OR3169-25G
OR3169-5G
APOSOR3169-1GEA
622
CZK
APOSOR3169-1G
APOSOR3169-25G
APOSOR3169-5G
5'-Bromo-2'-chloroacetophenone 99%
5'-Bromo-2'-chloroacetophenone
Vzorec:
C₈H₆BrClO MW: 233,4900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683851 CAS číslo: 105884-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...