(S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
Dodavatel: Apollo Scientific
Synonyms:
(S)-3-((Benzyloxy)carbonyl)oxazolidine-4-carboxylic acid, (S)-CBZ-Oxaproline
OR316041-1G
OR316041-5G
APOSOR316041-1GEA
6550
CZK
APOSOR316041-1G
APOSOR316041-5G
(S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
(S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
Vzorec:
C₁₂H₁₃NO₅ MW: 251,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00274193 CAS číslo: 97534-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...