Tisk…

L-Norvalinol

Dodavatel: Apollo Scientific

Synonyms: (S)-2-Aminopentan-1-ol

OR316038-5G
APOSOR316038-5GEA 3060 CZK
APOSOR316038-5G
L-Norvalinol
L-Norvalinol
Vzorec: C₅H₁₃NO
MW: 103,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683229
CAS číslo: 22724-81-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR