N-Boc-L-prolinal 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(S)-1-Boc-2-Formylpyrrolidine
OR316013-25G
APOSOR316013-25GEA
1700
CZK
APOSOR316013-25G
N-Boc-L-prolinal 97%
N-Boc-L-prolinal
Vzorec:
C₁₀H₁₇NO₃ MW: 199,25 g/mol Bod varu: 211 °C Hustota: 1,063 g/cm³ (25 °C) Bod vzplanutí: 57,2 °C Storage Temperature: Freezer |
MDL Number:
MFCD00274186 CAS číslo: 69610-41-9 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...