N-Boc-L-cyclohexylglycinol
Dodavatel: Apollo Scientific
Synonyms:
(S)-tert-Butyl (1-cyclohexyl-2-hydroxyethyl)carbamate
OR316005-5G
APOSOR316005-5GEA
4700
CZK
APOSOR316005-5G
N-Boc-L-cyclohexylglycinol
N-Boc-L-cyclohexylglycinol
Vzorec:
C₁₃H₂₅NO₃ MW: 243,3400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04112591 CAS číslo: 107202-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...