Tisk…

N-ɑ-Boc-L-tryptophanol 98%

Dodavatel: Apollo Scientific

Synonyms: (S)-2-(tert-Butoxycarbonyl)amino-3-(3-indolyl)propanol, N-ɑ-(tert-Butoxycarbonyl)-L-tryptophanol, (S)-2-(Boc-amino)-3-(3-indolyl)propanol, Boc-Tryptophanol, N-Boc-L-tryptophanol

OR316000-25G
APOSOR316000-25GEA 4140 CZK
APOSOR316000-25G
N-ɑ-Boc-L-tryptophanol 98%
N-alpha-Boc-L-tryptophanol
Vzorec: C₁₆H₂₂N₂O₃
MW: 290,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235953
CAS číslo: 82689-19-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR