Tisk…

3-Amino-2-naphthoic acid 97%

Dodavatel: Apollo Scientific

OR315769-5G OR315769-25G
APOSOR315769-5GEA 1740 CZK
APOSOR315769-5G APOSOR315769-25G
3-Amino-2-naphthoic acid 97%
3-Amino-2-naphthoic acid
Vzorec: C₁₁H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00004115
CAS číslo: 5959-52-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR