4,4'-Bis(chloromethyl)biphenyl
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4,4-Bis(chloromethyl)-1,1-biphenyl, 4,4'-Bis(chloromethyl)-1,1-biphenyl
OR315763-500G
APOSOR315763-500GEA
2820
CZK
APOSOR315763-500G
4,4'-Bis(chloromethyl)biphenyl
4,4'-Bis(chloromethyl)biphenyl
Vzorec:
C₁₄H₁₂Cl₂ MW: 251.15 g/mol Bod tání: 126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674019 CAS číslo: 1667-10-3 EINECS: 216-784-4 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...