Tisk…

1,2-Bis(diphenylphosphino)ethane

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

OR315704-100G OR315704-25G
APOSOR315704-100GEA 953 CZK
APOSOR315704-100G APOSOR315704-25G
1,2-Bis(diphenylphosphino)ethane
1,2-Bis(diphenylphosphino)ethane
Vzorec: C₂₆H₂₄P₂
MW: 398.42
Bod tání: 137…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS číslo: 1663-45-2
EINECS: 216-769-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR