Tisk…

Thiomorpholine-2-carboxylic acid

Dodavatel: Apollo Scientific

OR315576-1G OR315576-50MG OR315576-250MG OR315576-500MG OR315576-5G
APOSOR315576-1GEA 13030 CZK
APOSOR315576-1G APOSOR315576-50MG APOSOR315576-250MG APOSOR315576-500MG APOSOR315576-5G
Thiomorpholine-2-carboxylic acid
Thiomorpholine-2-carboxylic acid
Vzorec: C₅H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD09864920
CAS číslo: 134676-16-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR