Tisk…

8-Bromo-2-chloroquinazoline 97%

Dodavatel: Apollo Scientific

OR315552-5G
APOSOR315552-5GEA 3440 CZK
APOSOR315552-5G
8-Bromo-2-chloroquinazoline 97%
8-Bromo-2-chloroquinazoline
Vzorec: C₈H₄BrClN₂
MW: 243,4900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11043845
CAS číslo: 956100-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR