Tisk…

1-Bromo-2,2-dimethoxypropane 97%

Dodavatel: Apollo Scientific

OR315506-25G
APOSOR315506-25GEA 2150 CZK
APOSOR315506-25G
1-Bromo-2,2-dimethoxypropane 97%
1-Bromo-2,2-dimethoxypropane
Vzorec: BrH₂CC(OCH₃)₂CH₃
MW: 183,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000208
CAS číslo: 126-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR