Tisk…

Trioctylphosphine 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: TOP, Tri-n-octylphosphine

OR315487-100ML OR315487-25ML OR315487-500ML
APOSOR315487-100MLEA 2780 CZK
APOSOR315487-100ML APOSOR315487-25ML APOSOR315487-500ML
Trioctylphosphine 98%
Trioctylphosphine
Vzorec: C₂₄H₅₁P
Bod varu: 175 °C/0,3mmHg
Hustota: 0,831 g/cm³ (25 °C)
Bod vzplanutí: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00015298
CAS číslo: 4731-53-7
UN: 1760
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR