3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride 95+%
Dodavatel: Apollo Scientific
OR315411-1G
OR315411-250MG
OR315411-5G
APOSOR315411-1GEA
9200
CZK
APOSOR315411-1G
APOSOR315411-250MG
APOSOR315411-5G
3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride 95+%
3-Methyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride
Vzorec:
C7H16Cl2N2 MW: 199,1213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18083726 CAS číslo: 52407-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...