Tisk…

2-Bromo-9,9'-spirobi[9H-fluorene] 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Bromo-9,9'-spirobi[fluorene]

OR315380-25G
APOSOR315380-25GEA 2030 CZK
APOSOR315380-25G
2-Bromo-9,9'-spirobi[9H-fluorene] 97%
2-Bromo-9,9'-spirobi[9H-fluorene]
Vzorec: C₂₅H₁₅Br
MW: 395,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704217
CAS číslo: 171408-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR