Tisk…

5'-m-Terphenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: [1,1':3',1''-Terphenyl]-5'-ylboronic acid

OR315368-25G OR315368-100G
APOSOR315368-25GEA 3540 CZK
APOSOR315368-25G APOSOR315368-100G
5'-m-Terphenylboronic acid 98%
5'-m-Terphenylboronic acid
Vzorec: C₁₈H₁₅BO₂
MW: 274,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953491
CAS číslo: 128388-54-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR