Tisk…

Ethyl-5-cyanoindole-2-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 5-cyanoindole-2-carboxylate

OR315362-1G OR315362-5G
APOSOR315362-1GEA 2730 CZK
APOSOR315362-1G APOSOR315362-5G
Ethyl-5-cyanoindole-2-carboxylate
Ethyl-5-cyanoindole-2-carboxylate
Vzorec: C₁₂H₁₀N₂O₂
MW: 214.2200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04966999
CAS číslo: 105191-13-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR